This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03720394_m1
See other FGF13 GE Assays ›
Gene Symbol:
fgf13
Gene Name:
fibroblast growth factor 13
Gene Aliases:
fgf13.1, fgf2, fhf2
Chromosome Location:
Chr.8: 48576270 - 48764062 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03720394_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: fgf13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001079101.1 NP_001072569.1 3 - 4 533 79
XM_012968624.2 XP_012824078.1 4 - 5 663 79
XM_012968625.2 XP_012824079.1 4 - 5 707 79
XM_012968626.1 XP_012824080.1 2 - 3 1049 79
XM_012968627.2 XP_012824081.1 2 - 3 209 79
GenBank mRNA BC122912.1 - 3 - 4 533 79
CU025138.1 - 3 - 4 477 79
FJ480179.1 - 3 - 4 322 79
FJ480180.1 - 3 - 4 253 79
FJ480181.1 - 2 - 3 154 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

growth factor intercellular signal molecule

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline