Assay Details
| Gene Symbol: mtss1.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079332.1 | NP_001072800.1 | 386 | 82 | ||
| GenBank mRNA | BC121400.1 | - | 386 | 82 | ||
| Gene Symbol: LOC108644453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018089653.1 | XP_017945142.1 | 2 - 3 | 1056 | 82 | |
More Information
Set Membership: |
|

mtss1.1
Genomic Map