This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03724372_m1
See other QKI GE Assays ›
Gene Symbol:
qki
Gene Name:
QKI, KH domain containing, RNA binding
Gene Aliases:
Xqua, qk, qk1, qki-a, qki-b, quaking
Chromosome Location:
Chr.5: 57951182 - 58049678 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03724372_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: qki
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001100198.1 NP_001093668.1 2 - 3 713 91
XM_012962554.2 XP_012818008.1 2 - 3 903 91
XM_012962555.2 XP_012818009.1 2 - 3 904 91
XM_012962556.2 XP_012818010.1 2 - 3 905 91
XM_012962557.2 XP_012818011.1 2 - 3 900 91
XM_012962558.2 XP_012818012.1 2 - 3 901 91
XM_012962559.2 XP_012818013.1 2 - 3 905 91
XM_012962560.2 XP_012818014.1 2 - 3 901 91
XM_012962561.2 XP_012818015.1 2 - 3 901 91
XM_012962562.2 XP_012818016.1 2 - 3 903 91
XM_018093738.1 XP_017949227.1 3 - 4 523 91
XM_018093739.1 XP_017949228.1 2 - 3 904 91
GenBank mRNA BC135440.1 - 2 - 3 713 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0