This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03727850_m1
See other ELAVL4 GE Assays ›
Gene Symbol:
elavl4
Gene Name:
ELAV like neuron-specific RNA binding protein 4
Gene Aliases:
HuD, elrD, elrD1, elrD2
Chromosome Location:
Chr.4: 85528902 - 85591531 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03727850_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: elavl4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001103055.1 NP_001096525.1 4 - 5 496 98
XM_012960487.2 XP_012815941.1 4 - 5 574 98
XM_012960488.2 XP_012815942.1 4 - 5 577 98
XM_012960489.1 XP_012815943.1 4 - 5 692 98
XM_012960490.2 XP_012815944.1 3 - 4 487 98
XM_012960491.1 XP_012815945.1 4 - 5 814 98
XM_012960492.2 XP_012815946.1 3 - 4 490 98
XM_012960493.2 XP_012815947.1 3 - 4 606 98
XM_012960494.2 XP_012815948.1 3 - 4 528 98
XM_012960495.1 XP_012815949.1 3 - 4 587 98
GenBank mRNA BC135736.1 - 4 - 5 496 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0