This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03731037_m1
See other EEF1AKMT1 GE Assays ›
Gene Symbol:
eef1akmt1
Gene Name:
EEF1A lysine methyltransferase 1
Gene Aliases:
n6amt2
Chromosome Location:
Chr.2: 155910079 - 155916262 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03731037_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eef1akmt1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113847.1 NP_001107319.1 2 - 3 235 110
XM_012956383.2 XP_012811837.1 2 - 3 290 110
XM_012956384.2 XP_012811838.1 2 - 3 252 110
XM_012956386.2 XP_012811840.1 2 - 3 305 110
XM_012956387.1 XP_012811841.1 2 - 3 186 110
GenBank mRNA BC155986.1 - 2 - 3 235 110
BC156004.1 - 2 - 3 235 110
BC171256.1 - 2 - 3 224 110

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline