This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03731974_m1
See other VEGFA GE Assays ›
Gene Symbol:
vegfa
Gene Name:
vascular endothelial growth factor A
Gene Aliases:
vefg, vegf, vegf-a, vegfa-a, vegfa-b, vpf
Chromosome Location:
Chr.Un|NW_016683752: 29048 - 65435 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03731974_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: vegfa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001114232.1 NP_001107704.1 3 - 4 696 82
XM_012962542.2 XP_012817996.1 3 - 4 759 82
XM_012962543.2 XP_012817997.1 3 - 4 759 82
XM_012962544.2 XP_012817998.1 3 - 4 757 82
XM_012962545.2 XP_012817999.1 3 - 4 757 82
XM_012962547.2 XP_012818001.1 3 - 4 755 82
XM_012962548.1 XP_012818002.1 2 - 3 252 82
XM_012962549.2 XP_012818003.1 3 - 4 758 82
XM_012962550.2 XP_012818004.1 3 - 4 757 82
GenBank mRNA BC158522.1 - 3 - 4 696 82
CR760656.2 - 3 - 4 695 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

growth factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline