This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03732492_m1
See other NEIL1 GE Assays ›
Gene Symbol:
neil1
Gene Name:
nei-like DNA glycosylase 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 85499803 - 85512354 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03732492_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: neil1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001122798.1 NP_001116270.1 6 - 7 912 67
XM_012965879.2 XP_012821333.1 6 - 7 1006 67
XM_012965880.2 XP_012821334.1 6 - 7 950 67
XM_012965882.2 XP_012821336.1 6 - 7 1002 67
GenBank mRNA BC135399.1 - 6 - 7 914 67
BC161437.1 - 6 - 7 912 67
CR761671.2 - 5 - 6 834 67
CT485721.1 - 5 - 6 896 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

DNA glycosylase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline