This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03732503_m1
See other GTF2F1 GE Assays ›
Gene Symbol:
gtf2f1
Gene Name:
general transcription factor IIF subunit 1
Gene Aliases:
rap74
Chromosome Location:
Chr.3: 129664200 - 129674051 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03732503_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gtf2f1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001122799.1 NP_001116271.1 7 - 8 1446 73
XM_012953480.2 XP_012808934.1 8 - 9 1087 73
XM_012953481.2 XP_012808935.1 8 - 9 987 73
XM_012953482.2 XP_012808936.1 8 - 9 976 73
XM_012953483.2 XP_012808937.1 8 - 9 947 73
XM_012953484.2 XP_012808938.1 8 - 9 1088 73
XM_018091908.1 XP_017947397.1 8 - 9 986 73
GenBank mRNA BC135789.1 - 7 - 8 1119 73
BC161337.1 - 7 - 8 1446 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

general transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0