This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03735210_m1
See other YTHDF3 GE Assays ›
Gene Symbol:
ythdf3
Gene Name:
YTH N6-methyladenosine RNA binding protein 3
Gene Aliases:
-
Chromosome Location:
Chr.6: 103000932 - 103015882 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03735210_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ythdf3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_012964301.2 XP_012819755.1 4 - 5 229 87
XM_012964302.2 XP_012819756.1 4 - 5 299 87
XM_012964303.2 XP_012819757.1 3 - 4 473 87
XM_012964304.2 XP_012819758.1 3 - 4 701 87
XM_012964305.2 XP_012819759.1 3 - 4 231 87
XM_012964306.2 XP_012819760.1 2 - 3 448 87
XM_012964307.2 XP_012819761.1 3 - 4 270 87
XM_018094547.1 XP_017950036.1 2 - 3 371 87
GenBank mRNA BC161192.1 - 3 - 4 509 87
CT025387.2 - 3 - 4 519 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0