Assay Details
| Gene Symbol: fbxo24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128050.1 | NP_001121522.1 | 6 - 7 | 1309 | 71 | |
| GenBank mRNA | BC166383.1 | - | 6 - 7 | 1309 | 71 | |
More Information
Set Membership: |
|

fbxo24
Genomic Map