This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03741624_m1
See other CBLL1 GE Assays ›
Gene Symbol:
cbll1
Gene Name:
Cbl proto-oncogene-like 1, E3 ubiquitin protein ligase
Gene Aliases:
hakai, rnf188
Chromosome Location:
Chr.3: 66649600 - 66660981 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03741624_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cbll1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130242.1 NP_001123714.1 4 - 5 369 83
XM_012958649.2 XP_012814103.1 4 - 5 518 83
XM_012958650.2 XP_012814104.1 4 - 5 515 83
XM_012958651.2 XP_012814105.1 4 - 5 508 83
XM_018091951.1 XP_017947440.1 4 - 5 521 83
XM_018091952.1 XP_017947441.1 4 - 5 518 83
GenBank mRNA BC167325.1 - 4 - 5 369 83
CT027884.2 - 4 - 5 370 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0