This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03745426_m1
See other SLC25A16 GE Assays ›
Gene Symbol:
slc25a16
Gene Name:
solute carrier family 25 member 16
Gene Aliases:
d10s105e, gda, gdc, hgt.1, hml7, ml7
Chromosome Location:
Chr.7: 30450397 - 30470170 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03745426_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: slc25a16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001142210.1 NP_001135682.1 7 - 8 669 74
XM_012966233.2 XP_012821687.1 7 - 8 999 74
XM_012966234.2 XP_012821688.1 7 - 8 877 74
XM_012966235.2 XP_012821689.1 7 - 8 882 74
GenBank mRNA BC168609.1 - 7 - 8 669 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

mitochondrial carrier protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0