This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03745905_m1
See other FGF14 GE Assays ›
Gene Symbol:
fgf14
Gene Name:
fibroblast growth factor 14
Gene Aliases:
fhf4, sca27
Chromosome Location:
Chr.2: 116275879 - 116671998 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03745905_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: fgf14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001142824.1 NP_001136296.1 2 - 3 299 68
XM_018091121.1 XP_017946610.1 2 - 3 1202 68
XM_018091122.1 XP_017946611.1 3 - 4 640 68
GenBank mRNA FJ428270.1 - 2 - 3 299 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

growth factor intercellular signal molecule

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline