This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03746230_m1
See other SPIB GE Assays ›
Gene Symbol:
spib
Gene Name:
Spi-B transcription factor
Gene Aliases:
spi-b, spib-a, spib-b
Chromosome Location:
Chr.7: 109887872 - 109909638 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03746230_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: spib
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001145984.1 NP_001139456.1 4 - 5 360 62
XM_018095307.1 XP_017950796.1 4 - 5 451 62
GenBank mRNA FJ644947.1 - 4 - 5 360 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

winged helix/forkhead transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0