Assay Details
| Gene Symbol: mgst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011245.1 | NP_001011245.1 | 352 | 70 | ||
| GenBank mRNA | BC087821.1 | - | 352 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mgst1| Gene Symbol: mgst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011245.1 | NP_001011245.1 | 352 | 70 | ||
| GenBank mRNA | BC087821.1 | - | 352 | 70 | ||
Set Membership: |
|