This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Zm04038456_g1
See other LOC100280502 GE Assays ›
Gene Symbol:
LOC100280502
Gene Name:
SNARE-interacting protein KEULE
Gene Aliases:
ZEAMMB73_418140
Chromosome Location:
-
Species:
Zea mays
Species Specific ID (Flybase ID):
-
Assay ID Zm04038456_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100280502
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001153422.1 NP_001146894.1 1656 100
GenBank mRNA BT065394.1 - 1656 100
EU952152.1 - 1563 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Zea mays

Panther Classification:

Molecular Function -

membrane trafficking regulatory protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0