This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Zm04058212_g1
See other MLO8 GE Assays ›
Gene Symbol:
mlo8
Gene Name:
barley mlo defense gene homolog8
Gene Aliases:
-Mlo8, ZEAMMB73_016515, Zm-Mlo8
Chromosome Location:
-
Species:
Zea mays
Species Specific ID (Flybase ID):
-
Assay ID Zm04058212_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mlo8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001111701.1 NP_001105171.1 1207 70
GenBank mRNA AY029319.1 - 1207 70
AY104078.1 - 1207 70
BT043122.1 - 1386 70
BT056039.1 - 1279 70
EU971076.1 - 1253 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Zea mays

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0