This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Zm04059475_g1
See other MLO3 GE Assays ›
Gene Symbol:
mlo3
Gene Name:
barley mlo defense gene homolog3
Gene Aliases:
-Mlo3, Zm-Mlo3
Chromosome Location:
-
Species:
Zea mays
Species Specific ID (Flybase ID):
-
Assay ID Zm04059475_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mlo3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001112057.1 NP_001105527.1 253 61
GenBank mRNA AY029314.1 - 253 61
AY108219.1 - 237 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Zea mays

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0