man

Test

Viewing profile as user:
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Main Menu
Admin
Main Menu
ULS Inventory Manager
Products

CAS RN 1184609-55-9

CH3NSOOBr

CAS RN 1184609-55-9

IUPAC Name: 1-(2-bromobenzenesulfonyl)-3-methylpiperidine
Synonyms: 1-(2-bromobenzenesulfonyl)-3-methylpiperidine
Molecular Weight (g/mol): 318.23
Molecular Formula: C12H16BrNO2S
InChi Key: DHHOXSFLPKIJME-UHFFFAOYNA-N
SMILES: CC1CCCN(C1)S(=O)(=O)C1=CC=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results