CAS RN 14003-11-3
CAS RN 14003-11-3
IUPAC Name:
5-methylfuran-2-carbonyl chloride
Synonyms:
5-methylfuran-2-carbonyl chloride
Molecular Weight (g/mol):
144.55
Molecular Formula:
C6H5ClO2
InChi Key:
ZURUVZFDEVKNCE-UHFFFAOYSA-N
SMILES:
CC1=CC=C(O1)C(Cl)=O
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich 5-Methylfuran-2-carbonyl chloride
Sigma Aldrich 5-Methylfuran-2-carbonyl chloride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.