CAS RN 1451994-10-7
CAS RN 1451994-10-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ VU 0467485
Potent and selective positive allosteric modulator of M4 receptors; orally bioavailable and brain penetrant