CAS RN 1632296-13-9
CAS RN 1632296-13-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Caffeine Orange

Selective, BODIPY-based fluorescent caffeine chemosensor