accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 298-18-0

IUPAC Name: 2,2'-bioxirane
Synonyms: diepoxybutane bioxirane dioxybutadiene butane diepoxide 1,2:3,4-diepoxybutane DL butadiene dioxide 1,1'-bi[ethylene oxide] 1,3-butadiene diepoxide 1,2:3,4-butadiene diepoxide butadiendioxyd
Molecular Weight (g/mol): 86.09
Molecular Formula: C4H6O2
InChi Key: ZFIVKAOQEXOYFY-UHFFFAOYNA-N
SMILES: C1OC1C1CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results