accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4630-62-0

CAS RN 4630-62-0

IUPAC Name: 2-(hydroxymethyl)-6-phenoxyoxane-3,4,5-triol
Synonyms: β-D-glucopyranoside, phenyl
Molecular Weight (g/mol): 256.25
Molecular Formula: C12H16O6
InChi Key: NEZJDVYDSZTRFS-UHFFFAOYNA-N
SMILES: OCC1OC(OC2=CC=CC=C2)C(O)C(O)C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)