accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 514188-72-8

H3CSBr

CAS RN 514188-72-8

IUPAC Name: 2-bromo-5-decylthiophene
Synonyms: 2-bromo-5-decylthiophene
Molecular Weight (g/mol): 303.3
Molecular Formula: C14H23BrS
InChi Key: BLGUNQPKOOEOHU-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC1=CC=C(Br)S1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results