accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5470-34-8

CAS RN 5470-34-8

IUPAC Name: N-(4-methoxyphenyl)formamide
Synonyms: p-methoxyformanilde
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: SXEVZVMJNXOXIJ-UHFFFAOYSA-N
SMILES: COC1=CC=C(NC=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results