CAS RN 624741-47-5
CAS RN 624741-47-5
IUPAC Name:
3-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Synonyms:
3-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Molecular Weight (g/mol):
247.08
Molecular Formula:
C13H15BFNO2
InChi Key:
HIMZNJDDVIROEC-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=C(C=CC=C1F)C#N
1
–
1
of
1
results
1
6-Cyano-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
CAS: 624741-47-5 Molecular Formula: C13H15BFNO2 Molecular Weight (g/mol): 247.076 MDL Number: MFCD18729920 InChI Key: HIMZNJDDVIROEC-UHFFFAOYSA-N Synonym: 3-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile,3-fluoro-2-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile,3-fluoro-2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile,2-cyano-6-fluorophenyl boronic acid pinacol ester PubChem CID: 22240104 IUPAC Name: 3-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile SMILES: B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC=C2F)C#N