CAS RN 625437-46-9
CAS RN 625437-46-9
IUPAC Name:
N-[(4-fluorophenyl)methyl]cyclopropanamine
Synonyms:
N-[(4-fluorophenyl)methyl]cyclopropanamine
Molecular Weight (g/mol):
165.21
Molecular Formula:
C10H12FN
InChi Key:
YKNFXOCMGURJMK-UHFFFAOYSA-N
SMILES:
FC1=CC=C(CNC2CC2)C=C1
1
–
1
of
1
results
1
N-Cyclopropyl-4-fluorobenzylamine, 97%, Thermo Scientific™
CAS: 625437-46-9 Molecular Formula: C10H12FN Molecular Weight (g/mol): 165.21 MDL Number: MFCD07330007 InChI Key: YKNFXOCMGURJMK-UHFFFAOYSA-N Synonym: n-4-fluorobenzyl cyclopropanamine,cyclopropyl-4-fluoro-benzyl-amine,n-4-fluorophenyl methyl cyclopropanamine,cyclopropyl-4-fluorobenzyl amine,benzenemethanamine, n-cyclopropyl-4-fluoro,n-cyclopropyl-p-fluorobenzylamine,acmc-209n4r,n-cyclopropyl-4-fluorobenzylamine,n-4-fluorobenzyl cyclopropanamine #,n-4-fluorophenyl methyl cyclopropylamine PubChem CID: 573031 IUPAC Name: N-[(4-fluorophenyl)methyl]cyclopropanamine SMILES: FC1=CC=C(CNC2CC2)C=C1