CAS RN 62673-69-2
CAS RN 62673-69-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ GSK-3β Inhibitor VI
The GSK-3β Inhibitor VI, also referenced under CAS 62673-69-2, controls the biological activity of GSK-3β. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.