CAS RN 63857-91-0
CAS RN 63857-91-0
IUPAC Name:
2-(4-methylphenyl)-1H-imidazole-4,5-dicarbonitrile
Synonyms:
2-(4-methylphenyl)-1H-imidazole-4,5-dicarbonitrile
Molecular Weight (g/mol):
208.22
Molecular Formula:
C12H8N4
InChi Key:
BMTQOJUWHGWRPD-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)C1=NC(C#N)=C(N1)C#N
1
–
1
of
1
results
1
4,5-Dicyano-2-(4-methylphenyl)imidazole, 97%, Thermo Scientific™
CAS: 63857-91-0 Molecular Formula: C12H8N4 Molecular Weight (g/mol): 208.224 MDL Number: MFCD20265230 InChI Key: BMTQOJUWHGWRPD-UHFFFAOYSA-N Synonym: 4,5-dicyano-2-4-methylphenyl imidazole,2-p-tolyl-1h-imidazole-4,5-dicarbonitrile,2-4-methylphenyl-1h-imidazole-4,5-dicarbonitrile,4,5-dicyano-2-p-tolyl imidazole PubChem CID: 21798437 IUPAC Name: 2-(4-methylphenyl)-1H-imidazole-4,5-dicarbonitrile SMILES: CC1=CC=C(C=C1)C2=NC(=C(N2)C#N)C#N