man

Test

Viewing profile as user:
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Main Menu
Admin
Main Menu
ULS Inventory Manager
Products

CAS RN 639052-78-2

CAS RN 639052-78-2

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results