man

Test

Viewing profile as user:
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Main Menu
Admin
Main Menu
ULS Inventory Manager
Products

CAS RN 6590-93-8

ClClNCS

CAS RN 6590-93-8

IUPAC Name: 1,3-dichloro-5-isothiocyanatobenzene
Synonyms: 1,3-dichloro-5-isothiocyanatobenzene
Molecular Weight (g/mol): 204.07
Molecular Formula: C7H3Cl2NS
InChi Key: JLEMKZDHFGCHLO-UHFFFAOYSA-N
SMILES: ClC1=CC(=CC(Cl)=C1)N=C=S
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results