man

Test

Viewing profile as user:
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Main Menu
Admin
Main Menu
ULS Inventory Manager
Products

CAS RN 76972-35-5

CH3NNCl

CAS RN 76972-35-5

IUPAC Name: 1-chloro-4-(4-methylphenyl)phthalazine
Synonyms: 1-chloro-4-(4-methylphenyl)phthalazine
Molecular Weight (g/mol): 254.72
Molecular Formula: C15H11ClN2
InChi Key: SBSCJEDEQMLKJY-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=C2C=CC=CC2=C(Cl)N=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results