Thermo Scientific Spectroscopy Solutions for Education
education-spectroscopy-d education-spectroscopy-m
Overview
Engage and Motivate Students
Whether you are teaching introductory chemistry, quantitative analysis, organic chemistry or doing research with undergraduates, our instruments set the benchmark for high performance, affordability and ease of use. Dedicated science teachers know that hands-on work with high-quality instrumentation like the Thermo Scientific™ UV-visible and infrared spectrometers engages and energizes students.
"It used to be a drill session. Now you see the excitement as students grasp concepts with practical applications.”
— Sally Mitchell, ACS™ 2009 Chemistry Teacher of the Year.
Featured Product
spectronic-200-vis
SPECTRONIC 200 Visible Spectrophotometer
Make Learning Sample Analysis a Breeze
Get students excited about science with the Thermo Scientific™ SPECTRONIC™ 200 Visible Spectrophotometer. Based on decades of experience supporting the legendary "SPEC 20," the SPECTRONIC 200 instrument offers new technology and design features that will make your job easier, too!
Buy Now
Easy to Set Up:
  • No computer required — just plug it in and you're ready to measure
Easy to Use:
  • Familiar navigation and on-screen menus mean zero learning curve
  • Large, bright, color LCD screen adjusts for students of all heights
  • Convenient measurement wavelength selection with a rotatable knob or push-buttons
  • Standard sample compartment for 10mm square cuvettes and test tubes up to 25mm diameter
  • Room light resistant — run tall test tubes with the lid open
  • Cuvette racks on both sides of sample compartment help keep track of your samples
  • SPECTRONIC 20D+ emulation mode provides seamless integration into existing protocols
Easy to Maintain and Store:
  • Sample compartment lifts out for easy cleaning
  • Polymer construction resists corrosion from chemicals
  • Compact size saves space in the lab and when not in use can be stored on a book shelf
  • No motorized parts, so there's nothing to wear out
  More
Products
nanodrop-uv-vis
NanoDrop 2000 UV-Vis Spectrophotometer
Robust and reliable, the Thermo Scientific NanoDrop 2000 microvolume spectrophotometer can serve as an important tool in laboratories teaching molecular biology techniques such as cloning, plasmid preps, DNA and RNA purification and PCR.
Measure protein concentration in cell extracts using dye-based assays (e.g. BCA, Bradford, Lowry) and purified protein samples using absorbance at 280nm (A280). Determine bacteria or cell growth rates using OD600 measurements. Evaluate your plasmid mini preps for DNA concentration and purity. With the NanoDrop 2000 intuitive software, students can quickly learn to analyze their samples using a portable, convenient and accurate spectrophotometer.
Try Before You Buy
Buy Now
Benefits include:
  • Direct, easy measurements in less than 5 seconds — just pipette 1µL of sample, measure and wipe clean
  • Full spectral data output 190-840nm
  • Measure DNA, RNA (A260) and Protein (A280) concentrations and evaluate sample purity (A260/A280 and A260/A230 ratios)
  • Measure OD600 to monitor cell culture growth
  • Large concentration range (e.g. 2 ng/µL — 15,000 ng/µL for dsDNA samples); no dilutions required
  • Pre-configured software methods for common applications such as Nucleic Acid A260, Protein A280, Bradford, Lowry and more
  • Low-cost operation — no cuvettes to break, no consumables
  • Easy to maintain, easy to store
  •  
      More
    spectronic-200-vis
    SPECTRONIC 200 Visible Spectrophotometer
    Make Learning Sample Analysis a Breeze
    Get students excited about science with the Thermo Scientific™ SPECTRONIC™ 200 Visible Spectrophotometer. Based on decades of experience supporting the legendary "SPEC 20," the SPECTRONIC 200 instrument offers new technology and design features that will make your job easier, too!
    Buy Now
    Easy to Set Up:
    • No computer required — just plug it in and you're ready to measure
    Easy to Use:
    • Familiar navigation and on-screen menus mean zero learning curve
    • Large, bright, color LCD screen adjusts for students of all heights
    • Convenient measurement wavelength selection with a rotatable knob or push-buttons
    • Standard sample compartment for 10mm square cuvettes and test tubes up to 25mm diameter
    • Room light resistant — run tall test tubes with the lid open
    • Cuvette racks on both sides of sample compartment help keep track of your samples
    • SPECTRONIC 20D+ emulation mode provides seamless integration into existing protocols
    Easy to Maintain and Store:
    • Sample compartment lifts out for easy cleaning
    • Polymer construction resists corrosion from chemicals
    • Compact size saves space in the lab and when not in use can be stored on a book shelf
    • No motorized parts, so there's nothing to wear out
      More
    nicolet-is5-ft-ir
    Nicolet iS5 FT-IR Spectrometer
    Rugged and reliable, the Thermo Scientific Nicolet iS5 infrared spectrometer provides worry-free operation semester after semester. It's the perfect fit for classrooms and teaching laboratories — and now the value is better than ever.
    Buy Now
    Benefits include:
    • Small footprint
    • Superior performance
    • Flexible sampling
    • Intuitive software
    • Affordable price
    • Multi-year protection
    Reach your goal of engaged and educated students with the Thermo Scientific Nicolet iS5 FT-IR spectrometer.
      More
    genesys-10-s
    GENESYS 10S Visible Spectrophotometer
    The Thermo Scientific GENESYS 10S Vis instrument is ideal for instructional laboratories. Rugged construction and simple on-board controls combine with outstanding photometric performance to provide your students with the most accurate measurements.
    The GENESYS 10 Vis instrument goes beyond the basic visible spectrophotometer, offering precise measurements from 325nm to 1100nm and photometric linearity to 3 absorbance units.
    Buy Now
    Benefits include:
    • Easy transport and storage 
    • Operates as a stand-alone instrument or under computer control 
    • Acquire and display scanning, fixed wavelength, kinetics, or concentration data using embedded software and the graphical display 
    • Acquire data directly on a computer using VISIONlite 5™ software 
    • Save data to a USB memory device 
    • Increase sample throughput with the integrated six-cell changer 
    • Expand your capabilities with accessories for thermostatting and solid sample measurement
      More
    thermo-scientific-genesys-10s-spectrophotometer
    GENESYS 10S UV-Vis Spectrophotometer
    Utilize a high-intensity xenon lamp and dual-beam optical geometry to deliver unsurpassed data quality. Firing pulses of light only when the instrument is taking a measurement, the xenon lamp provides strong illumination from the UV to the near-IR region of the spectrum. You can count on the GENESYS 10S UV-Vis instrument for accurate and reliable results. The Thermo Scientific™ GENESYS™ 10 UV-Vis spectrophotometer offers an excellent value providing high performance, reliability, ease of use and a low cost of ownership.
    Buy Now
    Research Quality with Routine Simplicity
    • Scan speeds up to 4,200nm/minute
    • Depend on dual-beam optics for superior photometric accuracy
    • Acquire data from the UV to the near-IR
    • Choose from feature-rich local control or computer software options
    • Increase sample throughput with the integrated 6-cell changer
    • Save data to a USB memory device
    • Maintenance-Free Lamp
    • Save time with the instant-on xenon flash lamp
    • Perform accurate analysis over the entire wavelength range of 190-1100nm
    • Save money with the long-lifetime xenon flash lamp (guaranteed for 3 years)
      More
    thermo-scientific-pico-spin
    picoSpin 45 NMR Spectrometer
    The Thermo Scientific picoSpin 45 NMR spectrometer is the world's first desktop fourier transform-nuclear magnetic resonance (FT-NMR) spectrometer. Built to perform, this analytical tool is affordable and compact enough for any teaching, research or industrial lab.
    Download the picoSpin NMR Brochure (2.2MB)
    Buy Now
    Product highlights:
    • NMR analytical power at an economical price
    • No need for specialized facilities or training
    • Compact 45 MHz 1H pulsed FT-NMR spectrometer
    • Small sample volume (30 µL liquid samples)
    • High resolution at 60 ppb FWHM resolution or better
    • Small, lightweight and portable at 10.5 lbs. (4.8kg)
    • Fully programmable pulse sequencer
    • Includes 1-year license for MNova™ NMR software
    The picoSpin 45 NMR spectrometer is a true breakthrough in chemical instrumentation. Now you can afford a dedicated NMR spectrometer for your classroom or your research. Liquid samples are simply injected into an internal capillary via front-panel fittings, requiring only 20 to 30 µL of sample fluid to obtain a spectrum.
      More
    picoSpin NMR Awards
    Edison Award: Silver  R&D Award
    Mnova is a trademark of Mestrelab Research, Inc.