MOPS 10X Solution, Fisher BioReagents™
7732-18-5 1132-61-2 1310-73-2 3-(N-morpholino)propanesulfonic acid 3-morpholinopropane-1-sulfonic acid Premixed MOPS buffer solution (pH = 7.0) at 10X concentration. A low ionic strength buffer used for electrophoretic separation of formaldehyde denatured RNA.
Pricing and Availability
Buffer Solution, pH 10.00 (Certified), Fisher Chemical
6381-92-6 584-08-7 1310-58-3 7732-18-5 12228-88-5 Potassium Carbonate/Potassium Tetraborate/Potassium Hydroxide/Disodium EDTA Dihydrate.
Pricing and Availability
Tris Hydrochloride (Small White Flakes/Molecular Biology), Fisher BioReagents
1185-53-1 C4H11NO3·HCl H2NC(CH2OH)3·HCl Tris(hydroxymethyl)aminomethane Hydrochloride For electrophoresis, molecular biology, and cell culture
Pricing and Availability
BIS-TRIS Propane, JustPURE™, white powder, Fisher BioReagents
64431-96-5 C11H26N2O6 CH2[CH2NHC(CH2OH)3]2 1,3-Bis[tris(hydroxymethyl)methylamino]propane
Pricing and Availability
Tris-Acetate-EDTA, 10X Solution (Electrophoresis), Fisher BioReagents
64-19-7 7732-18-5 60-00-4 77-86-1 TAE For nucleic acid electrophoresis.
Pricing and Availability
Available on GSA/VA Contract for Federal Government customers only. Tris (Tromethamine) (Granular/USP), Fisher Chemical
77-86-1 (CH2OH)3CNH2 H2NC(CH2OH)3 Trishydroxymethylaminomethane Tris Buffer Tris THAM™ Trizma™
Pricing and Availability
Tris Base (White Crystals or Crystalline Powder/Molecular Biology), Fisher BioReagents
77-86-1 (CH2OH)3CNH2 H2NC(CH2OH)3 Trimethylol Aminomethane Tris(hydroxymethyl)aminomethane For molecular biology, tissue culture, and electrophoresis procedures.
Pricing and Availability
Gly-Gly, JustPURE™, white powder, Fisher BioReagents
556-50-3 C4H8N2O3 NH2CH2CONHCH2COOH Diglycine Glycyl-glycine
Pricing and Availability
Tris Hydrochloride, 1M Solution (pH 7.0, pH 7.5, and pH 8.0/Mol. Biol.), Fisher BioReagents
77-86-1 1185-53-1 7732-18-5 Tris-HCl For molecular biology, tissue culture, and electrophoresis procedures.
Pricing and Availability
Available on GSA/VA Contract for Federal Government customers only. PBS, 1X Powder Concentrate (White Granular Powder), Fisher BioReagents
7647-14-5 7558-79-4 7778-77-0 7447-40-7 PBS Commonly used buffer.
Pricing and Availability
Buffer Salt, pH 7.41 (Certified), Fisher Chemical
7558-79-4 7778-77-0 Potassium Phosphate Monobasic/Sodium Phosphate Dibasic Each packet contains sufficient material to prepare 1 liter of solution with a pH of 7.41 ±0.04 at 25°C.
Pricing and Availability
Buffer Concentrate, pH 10.00 (Certified), Fisher Chemical
7447-40-7 1310-73-2 139-33-3 7732-18-5 10043-35-3 Boric Acid/Potassium Chloride/Sodium Hydroxide.
Pricing and Availability
Tris-Borate-EDTA, 10X Solution (Electrophoresis), Fisher BioReagents
10043-35-3 60-00-4 77-86-1 7732-18-5 TBE For nucleic acid electrophoresis.
Pricing and Availability
Tris-Glycine, 10X Solution (Electrophoresis), Fisher BioReagents™
7732-18-5 77-86-1 56-40-6 TG Used as a buffer in native polyacrylamide gel electrophoresis.
Pricing and Availability
Ammonia ISE Fill Solution, Fisher Scientific™
107-21-1 7783-90-6 12125-02-9 7732-18-5 Stable under normal conditions of use and storage.
Pricing and Availability
Available on GSA/VA Contract for Federal Government customers only. TBE Buffer Mix (Dry Powder Mix of Tris/Boric Acid/EDTA/Electrophoresis), Fisher BioReagents
77-86-1 (Tris) 6381-92-6 (EDTA) 10043-35-3 (Boric Acid) Tris-Boric Acid-EDTA Buffer Mix
Pricing and Availability
Tris(hydroxymethyl)aminomethane (Cryst./Certified ACS), Fisher Chemical
77-86-1 (CH2OH)3CNH2 H2NC(CH2OH)3 2-Amino-2-(hydroxymethyl)-1,3-propanediol Trimethylol Aminomethane Tris Buffer Tris Trizma™ Alkalimetric standard.
Pricing and Availability
Tris(hydroxymethyl)aminomethane (Crystalline/Laboratory), Fisher Chemical
77-86-1 (CH2OH)3CNH2 H2NC(CH2OH)3 THAM* 2-Amino-2-(hydroxymethyl)-1,3-Propanediol Trimethylol Aminomethane Trizma™
Pricing and Availability
One moment while we fetch your results  spinner