Balance Software
Mettler Toledo™ LabX™ Direct Balance Software Available on GSA/VA Contract for Federal Government customers only.
Increases the efficiency and accuracy of weighing applications. Fisher Scientific - Mettler Toledo LabX Direct Balance Software is an easy-to-use software designed to transfer data from one balance into an open application like an Excel™ spreadsheet.
Mettler Toledo™ Balance Printer Accessories: Printer Paper Available on GSA/VA Contract for Federal Government customers only.
Printer paper for use with Mettler Toledo™ balances.
Mettler Toledo™ Optional Balance Interfaces
Interface for connection of an Excellence Balance to an Ethernet network.
Mettler Toledo™ Antistatic Systems Available on GSA/VA Contract for Federal Government customers only.
U-shaped and Point Ionizer antistatic systems for discharging electrostatically charged samples prior to weighing. Mettler Toledo Antistatic Systems use a high voltage to generate positively and negatively charged ions which are attracted by the electrostatically charged object and neutralize the disruptive charge on the surface of the weighing sample. As a result, the forces that distort the weighing result also disappear.
Mettler Toledo™ Compact Anti-Static Kit Available on GSA/VA Contract for Federal Government customers only.
For XP, XSE and XPE analytical balances.
OHAUS™ Sample Pans for MB35 and MB45 Moisture Analyzers
Sample Pans for Ohaus MB series moisture analyzers.
Mettler Toledo™ SmartCal™ Moisture Balance Reference Substances
Verify the performance of your moisture analyzer in just 10 minutes. Mettler Toledo SmartCal Moisture Balance Reference Substances can be run like any other sample to fulfill demands of industry regulation and company audits at the same time.
Mettler Toledo™ Ribbon Cartridge for Mettler Toledo Printers
Accessory for LC-P45, LC-P43, HA-P43 Printers.
OHAUS™ Glass Fiber Pads for MB35 and MB45 Moisture Analyzers Available on GSA/VA Contract for Federal Government customers only.
Accessory for use with MB35 and MB45 Moisture Analyzers.
Mettler Toledo™ Density Kit for NewClassic ME Balances
Accessory for NewClassic ME 0.1g/0.01g balance models.
Mettler Toledo™ Balance Printer Accessories: Cables
Printer accessories for use with Mettler Toledo balances.
Mettler Toledo™ Terminal Cover For XS/XA Series Analytical Balance Available on GSA/VA Contract for Federal Government customers only.
Terminal Cover For XS/XA Series Analytical Balances.
Mettler Toledo™ Temperature Calibration Kits Available on GSA/VA Contract for Federal Government customers only.
For use with halogen moisture analyzers. Mettler Toledo Temperature Calibration Kits are used to adjust halogen drying units to ensure consistent results.
OHAUS™ MB200 Series Balance Accessories Available on GSA/VA Contract for Federal Government customers only.
For use with Ohaus MB200 Moisture Analyzers.
Sartorius™ Glass Fiber Filters
For use with all Sartorius MA Models
Mettler Toledo™ Protective Covers for for NewClassic ME Balances
Protective covers for NewClassic ME balances.
Mettler Toledo™ ErgoClips™ Tube for Weighing
For use with XP, XS, XA analytical and XP, XS micro balance.
OHAUS™ 1050mL Bowl for CS Compact Scale
For use with CS Series Compact Scales.
OHAUS™ Sample Pan Handler for MB35 and MB45 Moisture Analyzers
For sue with MB35 and MB45 Moisture Analyzers.
Mettler Toledo™ ErgoClips™ Small Basket For Weighing
For use with test tubes, centrifuge tubes or small plastic tubes of XP, XS, XA analytical and XP, XS micro balance.
Mettler Toledo™ Damping Element
For underneath marble pad
OHAUS™ Accessories for Ranger™ Industrial Compact Bench and Counting Scales
For use with Ranger Industrial Compact Bench and Counting Scales.
Mettler Toledo™ Weighing Pan Set For XS/XP Series Analytical Balance
Weighing Pan Set For XS/XP Series Analytical Balances.
Mettler Toledo™ ErgoClips™ Flask For Weighing
For use with XP, XS, XA analytical and XP, XS micro balance.
  spinner