Plastic Beakers
Fisherbrand™ Polypropylene Disposable Beakers
Highly resistant to strong acids, organic solvents and other common laboratory reagents
Thermo Scientific™ Nalgene™ Polypropylene Griffin Low-Form Plastic Beakers Available on GSA/VA Contract for Federal Government customers only.
Carry out precise and efficient pouring with Thermo Scientific™ Nalgene™ Polypropylene Griffin Low-Form Beakers, featuring a single-spout design.
Fisherbrand™ Tri-Cornered Polypropylene Beakers
Unaffected by commonly used acids, alkalis and solvents
Fisherbrand™ Lids for Fisherbrand Polystyrene Beakers with Pouring Lip
For use with Fisherbrand Polystyrene Beakers with Pouring Lip (14-955-112).
Fisherbrand™ Polystyrene Beakers, 5mL
Ideal for use as sample containers
Corning™ Graduated Disposable Polypropylene Beakers with Lids Available on GSA/VA Contract for Federal Government customers only.
Manufactured from polypropylene for excellent chemical resistance. Corning™ Graduated Disposable Polypropylene Beakers serve as a beaker or storage container.
Thermo Scientific™ Nalgene™ PMP Griffin Low-Form Plastic Beakers Available on GSA/VA Contract for Federal Government customers only.
Continuously use Thermo Scientific™ Nalgene™ PMP Griffin Low-Form Beakers at temperatures of 150°C with intermittent service to 175°C.
Fisherbrand™ Low-Form Polypropylene Beakers
Smooth, tapered design facilitates easy handling and pouring
Bel-Art™ Polypropylene Graduated Pitchers Available on GSA/VA Contract for Federal Government customers only.
Constructed of translucent polypropylene. SP Scienceware™ Polypropylene Graduated Pitchers are ideal for general laboratory use and offer well designed spouts and large handles.
Fisherbrand™ Graduated Polystyrene Beakers
Economical throwaway beakers make excellent sample containers
Thermo Scientific™ Nalgene™ Griffin Low-Form PFA Plastic Beakers Available on GSA/VA Contract for Federal Government customers only.
Handle temperatures from -270° to +250°C using sturdy and tough Thermo Scientific™ Nalgene™ Griffin Low-Form PFA Beakers, ideal for continual lab usage.
Corning™ Graduated Disposable Polypropylene Beakers Available on GSA/VA Contract for Federal Government customers only.
Manufactured from polypropylene for excellent chemical resistance. Corning™ Graduated Disposable Polypropylene Beakers serve as a beaker or storage container.
Thermo Scientific™ Nalgene™ Griffin Low-Form Plastic Beaker Variety Pack
A convenient and cost effective way to get started setting up a new lab, assembling student lab kits, or refreshing an existing lab. Seven sizes, from 50 to 1000mL.
Bel-Art™ SP Scienceware™ Large Volume Polypropylene Beaker Available on GSA/VA Contract for Federal Government customers only.
Resistant to chemicals. SP Scienceware™ Large Volume Polypropylene Beaker features flared rims and spouts for quick, easy pouring when large volumes are handled.
Thermo Scientific™ Nalgene™ Graduated HDPE Plastic Beakers with Handles Available on GSA/VA Contract for Federal Government customers only.
Safely handle acids, alkalis and hot solutions with these beakers, featuring an extra-heavy wall and sturdy rectangular handle for reliable support and dispensing control.
Thermo Scientific™ Nalgene™ PMP Graduated Plastic Beakers with Handles Available on GSA/VA Contract for Federal Government customers only.
Ensure resistance against acids, bases, aliphatic alcohols, aldehydes and esters with Thermo Scientific™ Nalgene™ PMP Graduated Beakers.
Vitlab™ Polypropylene Griffin Beakers Available on GSA/VA Contract for Federal Government customers only.
Manufactured with highly transparent polypropylene for optimum visibility. Vitlab™ Polypropylene Griffin Beakers are embossed with easily readable raised blue scale for accurate measurement.
Alfa Aesar™ PTFE Beaker Cover-Watch Glass
For use as a beaker cover or a watch glass
Bel-Art™ SP Scienceware™ TPX™ Graduated Pitchers Available on GSA/VA Contract for Federal Government customers only.
Offer well-designed spouts and large handles helpful for use with wet or dry ingredients
Corning™ Reusable Plastic Beakers with Handle and Spout Available on GSA/VA Contract for Federal Government customers only.
Transluscent polypropylene beakers printed with blue graduations
Eisco Plastic Beaker; Premium Polypropylene
These polypropylene beakers have excellent optical clarity, very good chemical resistance, and are autoclavable. There are raised graduations and they have tapered spouts.
Bel-Art™ SP Scienceware™ Graduated Low-Form Griffin Beakers Available on GSA/VA Contract for Federal Government customers only.
Rugged beakers have permanent, raised numeric graduations and no-drip pour spout for easy pouring
Eisco Polypropylene Beaker with Handle and Spout
These polypropylene graduated cylinders have a handle and spout, are made from a single piece and are autoclavable.
Fisherbrand™ Intermediate-Form Polypropylene Beakers with Handles
Chemical-resistant, with a convenient pouring spout.
Eisco Polypropylene Beaker; Set of 5; 50ml, 100ml, 250ml, 500ml, 1000ml
These polypropylene beakers have excellent optical clarity, very good chemical resistance, and are autoclavable. There are raised graduations and they have tapered spouts.
Bel-Art™ SP Scienceware™ Tall Form Graduated Pitchers - Polypropylene
Bel-Art SP Scienceware Tall Form Graduated Pitchers are available in a large selection of sizes, and ideal for general laboratory use. Feature well-designed spouts and large open handles for easy stacking.
  spinner