NMR Tube Caps
DWK Life Sciences Kimble™ Kontes™ NMR Sample Tube Caps Available on GSA/VA Contract for Federal Government customers only.
For Kontes™ 5 or 10mm NMR sample tubes
DWK Life Sciences Kimble™ Kontes™ NMR Polyethylene Sample Tube Pressure Caps for 5 and 10mm Tubes
Tight-fitting, polyethylene cap which stays in place even with a build-up of internal vaporization pressures
Bel-Art™ SP Scienceware™ Wilmad™ Disposable NMR Tube Caps, 5.0mm Dia. Available on GSA/VA Contract for Federal Government customers only.
Different colors help track samples based on various reference solvents.
Wilmad™ SP Scienceware™ Gas-Tight Rubber Septa NMR Tube Cap Available on GSA/VA Contract for Federal Government customers only.
Designed to maintain excellent glass to rubber contact, the Wilmad-LabGlass Gas-Tight Rubber Septa NMR Tube Cap maintains a gas-tight seal while allowing for injection of samples or reactants.
Wilmad-LabGlass™ SP Scienceware™ PTFE NMR Tube Caps Available on GSA/VA Contract for Federal Government customers only.
Machined to exact specifications for greater spinning stability Wilmad-LabGlass PTFE NMR Tube Caps have a smaller gyroradius, more homogeneous mass distribution, and better chemical resistance than disposable caps.
DWK Life Sciences Kimble™ Kontes™ Replacement PTFE/Silicone Septa
For Kontes brand threaded NMR tubes K897635-0800 and K897735-1300, and open-top caps.
Bel-Art™ SP Scienceware™ Wilmad™ Superior Pressure NMR Tube Caps, 5.0mm Dia., Translucent Available on GSA/VA Contract for Federal Government customers only.
Ideal for experiments that require an air-tight environment under vacuum or low pressure less than 1 bar.
Bel-Art™ SP Scienceware™ Wilmad™ Disposable NMR Tube Caps, 4.0mm Dia., Blue Available on GSA/VA Contract for Federal Government customers only.
Different colors help track samples based on various reference solvents.
Kinesis™ Omnifit™ O-rings
For small and large variable bore cap-style connectors and valves
Bel-Art™ SP Scienceware™ Wilmad™ Disposable NMR Tube Caps, 8.0mm Dia., Neutral Available on GSA/VA Contract for Federal Government customers only.
Different colors help track samples based on various reference solvents.
Bel-Art™ SP Scienceware™ Wilmad™ Disposable NMR Tube Caps, 10.0mm Dia., Red Available on GSA/VA Contract for Federal Government customers only.
Different colors help track samples based on various reference solvents.
Bel-Art™ SP Scienceware™ Wilmad™ Disposable NMR Tube Caps, 2.5mm Dia., Red Available on GSA/VA Contract for Federal Government customers only.
Different colors help track samples based on various reference solvents.
Bel-Art™ SP Scienceware™ Wilmad™ Disposable NMR Tube Caps, 1.7mm Dia., Red Available on GSA/VA Contract for Federal Government customers only.
Different colors help track samples based on various reference solvents.
GE Healthcare O Rings for EPDM Glass Tubes Available on GSA/VA Contract for Federal Government customers only.
Different sized O Rings for EPDM Glass Tubes
  spinner