Stopper Accessories
DWK Life Sciences Kimble™ Kontes™ Silicone Stopper Assembly, 3 Port
3-port platinum-cured stopper assembly and sanitary PVDF clamp provides sterile fluid transfer with no extractables as required in biotech/pharmaceutical applications.
  spinner