Glass Dilution Bottles
Thermo Scientific™ Remel™ Butterfield's Phosphate Buffer (pH 7.2 stock solution)
Formulation used in food, dairy and pharmaceutical testing by AOAC, APHA, FDA, USDA, and USP test methods.
Corning™ Milk Dilution Bottles Available on GSA/VA Contract for Federal Government customers only.
Narrow- or wide-mouth borosilicate glass bottles with or without graduations
DWK Life Sciences Kimble™ KIMAX™-35 Milk Dilution Bottles Available on GSA/VA Contract for Federal Government customers only.
Designed from requirements for milk dilution bottles given in APHA Standard Methods for the Examination of Dairy Products
Kimble™ Kontes™ Static Dilution Bottle with Minert™ Valve
For purge and trap with Mininert™ valve
  spinner