Environmental Sampling Bottle Closures
Thermo Scientific™ LDPE Foam-lined Polypropylene Caps for Bulk Separates
LDPE-Lined White Caps are for general-purpose HDPE bulk separate containers.
Thermo Scientific™ I-Chem™ Loose Septa for 24-414 Open Top Caps
Thermo Scientific™ Loose Septa are replacement septa for Thermo Scientific VOA vials and septa bottles with 24-414 open top caps.
Thermo Scientific™ I-Chem™ Open-Top Septa Caps, 24-414mm, bonded septa, bulk
Thermo Scientific™ Septa Caps include a bonded-in 0.045″ thick septa.
Thermo Scientific™ PTFE-Lined Closed Caps
PTFE-Lined Closed Caps are replacement caps for Thermo Scientific glass environmental sample bottles, jars, jugs and vials.
  spinner