Flask General Accessories
Corning™ Disposable Aseptic Transfer Caps Available on GSA/VA Contract for Federal Government customers only.
Designed to fit Corning Polycarbonate Erlenmeyer flasks. Corning™ Disposable Aseptic Transfer Caps have aseptic venting and liquid transfer ports to minimize the risk of culture contamination.
Corning™ Plastic Erlenmeyer Flask Disposable Aseptic Transfer Caps Available on GSA/VA Contract for Federal Government customers only.
Allows you to fill or empty the Corning 5L Erlenmeyer flask in a closed environment. Fluid is moved by positive or negative pressure via a vacuum pump to fill (apply a vacuum) or empty (pressurize) the cell culture vessel.
Corning™ GL45 Disposable Aseptic Transfer Cap Available on GSA/VA Contract for Federal Government customers only.
Reduces contamination and enhancse the productivity of cell culture operations when adding or removing medium and cells
CleanWear™ Tyvek™ Round Covers
Durable, form-fitting cover for small bowls, flasks or other consumables.
DWK Life Sciences Kimble™ Kontes™ Center Well for Incubation Flask Available on GSA/VA Contract for Federal Government customers only.
The top stopper for the Kontes™ sidearm incubation flask has an off-center hole for the insertion of the disposable polypropylene center well. The center well may be cut off when transferred to a liquid scintillation counting vial.
Corning™ Clamp for 125mL Erlenmeyer Flask
For use with 125mL Erlenmeyer flask, Corning LSE orbital shaker and Corning LSE low speed orbital shaker.
Fisherbrand™ Cell Washer
Single- or double-cell capacity
DWK Life Sciences Kimble™ Kontes™ Top Stopper for Incubation Flask Available on GSA/VA Contract for Federal Government customers only.
Fits both 10 and 25mL flasks. The rubber top stopper for the Kontes™ sidearm incubation flask provides a double seal and has an off-center hole for the insertion of the center well.
DWK Life Sciences Kimble™ Filter Flasks Spare Parts - Plastic Sidearm and Bushing
Replacement plastic Sidearm and Bushing for graduated filter flasks.
DWK Life Sciences Kimble™ Desiccator Parts: Neoprene Bushings
Replacement Part - Case of 10 Neoprene Bushings
Corning™ ProCulture Media Handling Fittings Available on GSA/VA Contract for Federal Government customers only.
Introduce medium aseptically into larger Spinner Flasks with vertical sidearms during cell culture
Corning™ ProCulture Gas Delivery Fittings Available on GSA/VA Contract for Federal Government customers only.
Used to provide gases into larger Spinner Flasks with vertical sidearms during cell culture
Corning™ ProCulture™ Gas/Media Handling Fittings For 6L Angled Sidearm Flasks Available on GSA/VA Contract for Federal Government customers only.
Includes PET insert with fluorocarbon O-ring and polypropylene-sealing cap
  spinner