Gas Washing Bottles
Kimble™ Kontes™ Guth Unitized™ Wash Bottle
One-handed sanitary manipulation
Bel-Art™ SP Scienceware™ Gas Washing Bottle with Fritware Dispersion Disc Available on GSA/VA Contract for Federal Government customers only.
Practical and economical graduated 250mL cylinder with a stable extended base for use in precipitating elements from gas
Kimble™ Kimble™Kontes™ Tall Form Gas Washing Bottle with Medium Length Joint and Kimflow™ Cylinder
KIMAX™ bottle used to wash or dry (scrub) gases of undesirable constituents.
Kimble™ Kontes™ Gas Washing Bottle
Tall form gas washing bottle with full-length joint with hooks contains a fritted cylinder disc for more uniform gas distribution throughout the absorbing material. Available in two porosities: extra coarse (170-220 micron with 40/50 standard taper joint) and coarse (40-60 micron with standard 29/42 taper joint).
  spinner