Glass Centrifuge Bottles
PYREX™ Glass Centrifuge Bottles Available on GSA/VA Contract for Federal Government customers only.
Permit concentration of sediment for decanting operations
DWK Life Sciences Kimble™ KIMAX™ Babcock Bottles for Cheese Testing Available on GSA/VA Contract for Federal Government customers only.
Paley centrifuge bottles feature large stoppered opening next to neck to accommodate both liquids and solids
DWK Life Sciences Kimble™ KIMAX™ Babcock Bottles for Ice Cream Testing Available on GSA/VA Contract for Federal Government customers only.
Scale is marked with permanent brown stain on neck.
DWK Life Sciences Kimble™ KIMAX™ Babcock Bottle for Gasoline Testing Available on GSA/VA Contract for Federal Government customers only.
For determination of unsaturated hydrocarbons in gasoline
  spinner