Round Glass Wide Mouth Bottles
Fisherbrand™ Graduated Glass Bottle Beakers with White Polypropylene PE Foam/PTFE Cap
Environmentally sensitive bottles help eliminate waste. Fisherbrand™ Clear Graduated Clear Graduated Boston Round Bottles with White Polypropylene PE Foam/PTFE Lined Cap ensures product integrity for long term storage.
Fisherbrand™ Clear Standard Wide Mouth Bottles with White Polypropylene PE Foam/PTFE Cap Available on GSA/VA Contract for Federal Government customers only.
Ideal general purpose bottle. Fisherbrand™ Clear Standard Wide Mouth Bottles with White Polypropylene PE Foam/PTFE Cap used for pharmaceutical, chemical, environmental, and food packaging and storing applications.
PYREX™ Round Wide-Mouth Media Storage Bottles Available on GSA/VA Contract for Federal Government customers only.
Designed for heavy-duty storage as well as mixing and sampling
Fisherbrand™ Amber Wide Mouth Packer Bottles with White Polypropylene PE Foam/PTFE Cap Available on GSA/VA Contract for Federal Government customers only.
Feature a wide opening, making them ideal general purpose bottles for solids. Fisherbrand™ Amber Wide-Mouth Packers with PTFE-Faced PE-Lined Caps are the classic vitamin bottles, and are often used in pharmaceutical, chemical, and environmental applications. The amber glass filters out ultraviolet wavelengths to protect light-sensitive solids.
Fisherbrand™ Custom Cleaned Amber Wide Mouth Packer Bottles
Ideal storage for light sensitive dry powders. Fisherbrand™ Custom Cleaned Amber Wide Mouth Packer Bottles are manufactured using amber soda glass.
Fisherbrand™ Graduated Glass Bottle Beakers with White Polypropylene Pulp/Vinyl Cap
Multipurpose containers for use with biological and pathological specimens. Fisherbrand™ Clear Graduated Glass Bottles with Polypropylene Pulp/Vinyl Lined Caps can also be used for storing industrial laboratory chemicals and reagents.
Qorpak™ Amber Wide Mouth Packer Bottles, Vacuum and Ionized with Thermoset F217 and PTFE Cap Available on GSA/VA Contract for Federal Government customers only.
Provides additional protection for light-sensitive contents by blocking UV rays. Qorpak™ Amber Wide Mouth Packer Bottles, Vacuum and Ionized with Thermoset F217 and PTFE Cap is useful for wide range of applications including pharmaceutical, chemical and environmental.
DWK Life Sciences Wheaton™ Glass Sample Bottles Available on GSA/VA Contract for Federal Government customers only.
Ideal for collecting, storing and exhibiting samples in the lab or field. Wheaton™ Snap Cap Sample Bottles are manufactured from Wheaton™ 800 soda lime clear glass that conforms to USP Type III requirements.
Qorpak™ Glass Bottle Beakers with Green Thermoset F217 and PTFE Caps
Can be used for mixing, measuring and storage
Qorpak™ Clear Straight Sided Round Bottles with Black Phenolic Pulp/Vinyl Cap Available on GSA/VA Contract for Federal Government customers only.
Provides content visibility. Qorpak™ Clear Straight Sided Round Bottles with Black Phenolic Pulp/Vinyl Cap are versatile multipurpose jars.
Fisherbrand™ Clear Standard Wide Mouth Bottles with White Polypropylene Pulp/Vinyl Lined Cap Available on GSA/VA Contract for Federal Government customers only.
General purpose bottle. Fisherbrand™ Clear Standard Wide Mouth Bottles with White Polypropylene Pulp/Vinyl Lined Cap are used for pharmaceutical, chemical, environmental, and food packaging and storing applications.
DWK Life Sciences Kimble™ Clear Glass AC Medium Rounds without Caps Available on GSA/VA Contract for Federal Government customers only.
Wide mouth design for efficient addition or removal of contents
Qorpak™ Amber Wide Mouth Packer Bottles with Green Thermoset F217 and PTFE Cap Available on GSA/VA Contract for Federal Government customers only.
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Glass Jug with Green Thermoset F217 and PTFE Cap are used in chemical and environmental applications.
Fisherbrand™ Graduated Glass Bottle Beakers with Black Phenolic PolyCone Cap
Multipurpose containers for use with biological and pathological specimens. Fisherbrand™ Graduated Glass Bottle Beakers with Black Phenolic PolyCone Cap can also be used for storing industrial laboratory chemicals and reagents.
Fisherbrand™ Amber Wide Mouth Packer Bottles with Black Polyseal Polyethylene Cone Cap Available on GSA/VA Contract for Federal Government customers only.
Ideal for storing dry materials. Fisherbrand™ Amber Wide Mouth Packer Bottles with Black Polyseal Polyethylene Cone Cap are often used in chemical and environmental applications.
DWK Life Sciences Wheaton™ HDPE Sample Bottles Available on GSA/VA Contract for Federal Government customers only.
Ideal for collecting, storing and exhibiting samples in the lab or field. Wheaton™ Snap Cap Sample Bottles are manufactured from Wheaton™ 800 soda lime clear glass that conforms to USP Type III requirements.
Qorpak™ Clear Graduated Medium Round Ultra Clean Bottles with Thermoset Caps
For use with biological and pathological specimens. Qorpak™ Clear Graduated Medium Round Ultra Clean Bottles are used for storing industrial laboratory chemicals and reagents.
Qorpak™ Graduated Medium Round Bottles, Vacuum and Ionized with Black Phenolic Pulp/Vinyl Caps
Odor-free and tasteless with no need to clean before initial use
DWK Life Sciences Wheaton™ Bottles Amber Wide-Mouth Packers with Polyvinyl-lined Closures Available on GSA/VA Contract for Federal Government customers only.
Amber glass for storing light-sensitive powders. Wheaton™ Bottles Amber Wide-Mouth Packers with Polyvinyl-lined Closures are available in several capacities.
Fisherbrand™ Clear Standard Wide Mouth Bottles without Cap Available on GSA/VA Contract for Federal Government customers only.
For pharmaceutical, chemical, environmental applications. Fisherbrand™ Clear Standard Wide Mouth Bottles without Cap are made of glass.
Qorpak™ Clear Straight Sided Round Bottles with Green Thermoset F217 and PTFE Caps
Designed without shoulders for maximum storage capacity
  spinner