Plastic Dilution Bottles
Thermo Scientific™ Nalgene™ Polysulfone Dilution Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Resist bases, weak acids and all aqueous solutions in applications with our clearest and most accurate dilution bottles.
Thermo Scientific™ Nalgene™ PPCO Dilution Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Easily view liquid levels with these translucent PPCO dilution bottles, which offer excellent chemical resistance to strong acids, alcohols and bases.
  spinner