Plastic Roller Bottles
Corning™ Disposable Roller Bottles Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically clear virgin polystyrene for excellent visibility and tissue culture-treated for optimal cell attachment. Corning™ Disposable Roller Bottles allow Initial scale-up of adherent cells with intermittent exposure of cells to the air and the media.
Corning™ Polystyrene Roller Bottles Available on GSA/VA Contract for Federal Government customers only.
Treated with patented Corning CellBIND™ surface treatment
Thermo Scientific™ Nunc™ PETG Roller Bottles Available on GSA/VA Contract for Federal Government customers only.
Increase cell expansion and product yield while eliminating the need to purchase additional production equipment or increase labor.
Corning™ Expanded-Surface Roller Bottles Available on GSA/VA Contract for Federal Government customers only.
Features a ribbed design that doubles the growth area without increasing bottle size. Corning™ Expanded Surface Roller Bottles one-piece seamless polystyrene construction prevents warping or bowing during use.
Greiner Bio-One™ CELLMASTER™ Polystyrene Filter Cap Roller Bottles – Short Form
For use in cell culture. Greiner Bio-One CELLMASTER™ Polystyrene Filter Cap Roller Bottles – Short Form provides cell adhesion through physical surface treatment.
Corning™ Roller Bottle with Easy Grip Cap Available on GSA/VA Contract for Federal Government customers only.
One piece seamless construction with printed graduations
Thermo Scientific™ Nunc™ Polystyrene Roller Bottles Available on GSA/VA Contract for Federal Government customers only.
Reliably grow cells with high reproducibility by using Thermo Scientific™ Nunc™ Polystyrene Roller Bottles.
Corning™ Roux Culture Bottles
For growing mass cultures and single or monolayer cultures. When bottle is laid flat, it will hold approximately one half of the stated capacity of solution.
Greiner Bio-One™ CELLMASTER™ Polystyrene Roller Bottles with Filter Cap
For use in cell culture. Greiner Bio-One CELLMASTER™ Polystyrene Roller Bottles with Filter Cap stable and transparent.
Greiner Bio-One™ CELLMASTER™ Polystyrene Roller Bottles - Short Form
For use in cell culture. Greiner Bio-One CELLMASTER™ Polystyrene Roller Bottles - Short Form is stable and transparent and comes with safety screw cap.
Corning™ CellBIND™ 850cm Squared Polystyrene Roller Bottle With Easy Grip Cap Available on GSA/VA Contract for Federal Government customers only.
Features large knurls designed for ergonomic handling.
Greiner Bio-One™ CELLMASTER™ Polystyrene Roller Bottles - Long Form
For use in cell culture. Greiner Bio-One CELLMASTER™ Polystyrene Roller Bottles - Long Form renders cell adhesion through physical surface treatment.
Greiner Bio-One™ CELLMASTER™ Polyethylene Terephthalate (PET) Roller Bottles - Short Form
For use in cell culture. Greiner Bio-One CELLMASTER™ Polyethylene Terephthalate (PET) Roller Bottles - Short Form is suitable for many adherent cell lines, is stable and gas permeable.
Greiner Bio-One™ CELLMASTER™ Polystyrene 5XL Roller Bottles - Long Form
For use in cell culture. Greiner Bio-One CELLMASTER™ Polystyrene 5XL Roller Bottles - Long Form renders cell adhesion through physical surface treatment.
Greiner Bio-One™ CELLMASTER™ Polystyrene Roller Bottles with Ribbed Surface – Short Form
For use in cell culture. Greiner Bio-One CELLMASTER™ Polystyrene Roller Bottles with Ribbed Surface – Short Form gives stability and transparency and features safety cap.
Greiner Bio-One™ CELLMASTER™ Glycolised Polyethylene Terephthalate (PETG) 2X Roller Bottles - Short Form
For use in cell culture. Greiner Bio-One CELLMASTER™ Glycolised Polyethylene Terephthalate (PETG) 2X Roller Bottles - Short Form is suitable for many adherent cell lines, is stable and gas permeable.
Greiner Bio-One™ CELLMASTER™ Polystyrene Filter Cap Roller Bottles - Long Form
For use in cell culture. Greiner Bio-One CELLMASTER™ Polystyrene Filter Cap Roller Bottles - Long Form renders cell adhesion through physical surface treatment.
Greiner Bio-One™ CELLMASTER™ Polystyrene 2.5X Roller Bottles - Short Form
For use in cell culture. Greiner Bio-One CELLMASTER™ Polystyrene 2.5X Roller Bottles - Short Form renders cell adhesion through physical surface treatment.
Greiner Bio-One™ CELLMASTER™ Polystyrene Filter Cap Roller Bottles - Long Form
For use in cell culture. Greiner Bio-One CELLMASTER™ Polystyrene Filter Cap Roller Bottles - Long Form renders cell adhesion through physical surface treatment.
BD Falcon™ Roller Bottles, Vented Cap
Compatible with manual and automated filling systems
  spinner