Round Plastic Narrow Mouth Bottles
Thermo Scientific™ Nalgene™ Boston Round Opaque White HDPE Bottles without Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Protect light-sensitive products with these chemically-resistant HDPE bottles, which meet the specifications of light-resistant containers per USP requirements.
Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Packaging Bottles with Closure: Sterile, Shrink-Wrapped Trays Available on GSA/VA Contract for Federal Government customers only.
Minimize biological and particulate contamination with sterile packaging bottles produced in a controlled environment and featuring chemical resistance.
Thermo Scientific™ Nalgene™ Narrow-Mouth Natural HDPE Packaging Bottles with Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Store, ship and package specialty chemicals, diagnostics, biologicals, reagents, adhesives and veterinary pharmaceuticals with these narrow-mouth HDPE bottles.
Thermo Scientific™ Nalgene™ Natural HDPE Diagnostic Bottles with Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Sample, store and ship small volume, valuable reagents. These leakproof bottles with polypropylene closures are used in test kits and many small-volume applications.
Thermo Scientific™ Nalgene™ Clear PETG Diagnostic Bottles with Closure: Sterile, Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Sample, store and ship small volume, valuable reagents with clear PETG diagnostic bottles that are ideal for use in many sterile, small-volume applications.
Thermo Scientific™ Nalgene™ Narrow-Mouth Opaque Amber HDPE Packaging Bottles with Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Store and ship light-sensitive reagents with these bottles, ideal for diagnostics, biologicals, specialty chemicals, adhesives and veterinary pharmaceuticals.
Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles with Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Safely store and ship light-sensitive, small-volume valuable reagents with these opaque amber HDPE diagnostic bottles, for use in test kits and many small-volume applications.
Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Lab Quality Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Store, ship and package liquids with these highly reliable, lab quality bottles, specifically designed for long-term demanding lab use.
Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Bottles with Closure: Shrink-Wrapped Trays Available on GSA/VA Contract for Federal Government customers only.
Easily handle these narrow-mouth HDPE bottles that are ready to feed into filling lines for various reagents, and conveniently packaged in shrink-wrap trays.
Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles with Closure: Nonsterile, Tray Pack Available on GSA/VA Contract for Federal Government customers only.
Perform many small volume applications with these safe and leakproof diagnostic bottles, ideal for use in test kits, sample storage and reagent shipping.
Thermo Scientific™ Nalgene™ Natural HDPE Diagnostic Bottles without Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Store and ship small volume, valuable reagents with Thermo Scientific™ Nalgene™ Natural HDPE Diagnostic Bottles without Closure.
Thermo Scientific™ Nalgene™ Boston Round Narrow-Mouth HDPE Bottles with Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Package and ship liquids with Thermo Scientific™ Nalgene™ Boston Round Narrow-Mouth HDPE Bottles with Closure, which are both durable and reliable.
Corning™ Costar™ Disposable Storage Bottles Available on GSA/VA Contract for Federal Government customers only.
Manufactured from clear polystyrene for strength and optimum visibility. Corning™ Costar™ Disposable Storage Bottles offer the ideal solution for storing media, buffers, and other aqueous solutions.
Thermo Scientific™ Nalgene™ Narrow-Mouth Polycarbonate Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Freeze samples effectively with impact-resistant Thermo Scientific™ Nalgene™ Narrow-Mouth Polycarbonate Bottles with Closure.
Thermo Scientific™ Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Store corrosive chemicals and distilled water with Thermo Scientific™ Nalgene™ Large Narrow-Mouth PPCO Bottles, which are resistant to most acids, bases and alcohols.
Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles without Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Store samples and ship small volumes of valuable reagents and light sensitive diagnostic samples with Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles without Closure.
Thermo Scientific™ Nalgene™ Narrow-Mouth Amber HDPE Lab Quality Bottles Available on GSA/VA Contract for Federal Government customers only.
Store, ship and package light-sensitive liquid using these narrow-mouth, amber HDPE bottles. They offer excellent chemical resistance to most acids, bases and alcohols.
Thermo Scientific™ Nalgene™ Narrow-Mouth Teflon™ PFA Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Get excellent resistance to temperature, chemicals and corrosion with the narrow-mouth bottles that among the most chemical- and corrosion-resistant containers available.
Thermo Scientific™ Nalgene™ Narrow-Mouth LDPE Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Carry out applications such as trace metal analysis with flexible, impact-and chemical-resistant Thermo Scientific™ Nalgene™ Narrow-Mouth LDPE Bottles with Closure.
Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles with Closure: Sterile, Tray-Packed Available on GSA/VA Contract for Federal Government customers only.
Perform many small volume packing applications with Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles with Closure.
Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles without Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Safely store and ship light-sensitive diagnostic samples and valuable reagents in small volumes with Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles.
Thermo Scientific™ Nalgene™ PETG Diagnostic Bottles with Closure: Sterile Available on GSA/VA Contract for Federal Government customers only.
Conduct sterile sampling, storage and shipment of reagents with these clear PETG bottles that include leakproof, lined white HDPE closures.
Thermo Scientific™ Nalgene™ Certified Low Particulate Narrow-Mouth HDPE Bottles with Closure: Lab Pack Available on GSA/VA Contract for Federal Government customers only.
Effectively store high-purity chemicals with these HDPE bottles, which are extremely low in trace metal content.
Thermo Scientific™ Nalgene™ Narrow-Mouth PPCO Bottles with Closure: Autoclavable Available on GSA/VA Contract for Federal Government customers only.
Perform lab applications requiring excellent chemical resistance and autoclaving with and without contents using Thermo Scientific™ Nalgene™ Narrow-Mouth PPCO Bottles with Closure.
Thermo Scientific™ Nalgene™ Teflon™ FEP Low Particulate/ Low Metals Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Optimize storage and sampling of active pharmaceutical ingredients and bulk intermediates by using these bottles, certified to particulate and trace metals levels.
Thermo Scientific™ Nalgene™ Boston Round Narrow-Mouth Natural PPCO Bottles with Closure: Bulk Pack Available on GSA/VA Contract for Federal Government customers only.
Ensure continued stock of Thermo Scientific™ Boston Round Narrow-Mouth Natural PPCO Bottles with Closure with this bulk-packed option.
Thermo Scientific™ Nalgene™ Narrow-Mouth Teflon™ FEP Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.
Conduct trace metal analysis, high- and low-temperature work and applications involving organic solvents or rigorous cleaning.
Thermo Scientific™ Nalgene™ Large Narrow-Mouth LDPE Bottles Available on GSA/VA Contract for Federal Government customers only.
Store chemicals and standards and collect and store distilled water. These translucent bottles offer flexibility, impact resistance and visibility of contents.
  spinner