Roller Bottles
Corning™ Expanded-Surface Roller Bottles Available on GSA/VA Contract for Federal Government customers only.
Features a ribbed design that doubles the growth area without increasing bottle size. Corning™ Expanded Surface Roller Bottles one-piece seamless polystyrene construction prevents warping or bowing during use.
Corning™ Disposable Roller Bottles Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically clear virgin polystyrene for excellent visibility and tissue culture-treated for optimal cell attachment. Corning™ Disposable Roller Bottles allow Initial scale-up of adherent cells with intermittent exposure of cells to the air and the media.
Corning™ Polystyrene Roller Bottles Available on GSA/VA Contract for Federal Government customers only.
Treated with patented Corning CellBIND™ surface treatment
Thermo Scientific™ Nunc™ PETG Roller Bottles Available on GSA/VA Contract for Federal Government customers only.
Increase cell expansion and product yield while eliminating the need to purchase additional production equipment or increase labor.
Corning™ Roller Bottle with Easy Grip Cap Available on GSA/VA Contract for Federal Government customers only.
One piece seamless construction with printed graduations
Thermo Scientific™ Nunc™ Polystyrene Roller Bottles Available on GSA/VA Contract for Federal Government customers only.
Reliably grow cells with high reproducibility by using Thermo Scientific™ Nunc™ Polystyrene Roller Bottles.
Corning™ CellBIND™ 850cm Squared Polystyrene Roller Bottle With Easy Grip Cap Available on GSA/VA Contract for Federal Government customers only.
Features large knurls designed for ergonomic handling.
Corning™ Roux Culture Bottles
For growing mass cultures and single or monolayer cultures. When bottle is laid flat, it will hold approximately one half of the stated capacity of solution.
PYREX™ Glass Roller Bottles Available on GSA/VA Contract for Federal Government customers only.
Withstand repeated wet or dry sterilization
DWK Life Sciences Wheaton™ Glass Roller Culture Vessels
Uniform quality and performance for higher cell yields
  spinner