Burettes
Thermo Scientific™ Nalgene™ Break-Resistant Self-Zeroing Automatic Burets Available on GSA/VA Contract for Federal Government customers only.
Ensure repeatable, accurate results with Thermo Scientific™ Nalgene™ Break-Resistant Self-Zeroing Automatic Burets (meets ASTM E287).
Thermo Scientific™ Nalgene™ Acrylic Burets with PMP Tip, TFE Stopcock and Plug Available on GSA/VA Contract for Federal Government customers only.
Accurately perform applications with Thermo Scientific™ Nalgene™ Acrylic Burets with PMP Tip, TFE Stopcock and Plug, which meet ASTM E287 standards.
Thermo Scientific™ Nalgene™ Self-Zeroing Buret Filler Kit Available on GSA/VA Contract for Federal Government customers only.
Save time and increase convenience. This filler kit converts any conventional glass or plastic 25mL or 50mL buret into an automatic-filling, self-zeroing buret.
Fisherbrand™ Castaloy™ Double-Buret Clamp
Holds two burets from micro to 100mL size
PYREX™ Class A Buret with Product Standard PTFE Stopcock Plug Available on GSA/VA Contract for Federal Government customers only.
Buret has colored markings, fine, sharp lines and large easy-to-read numbers
DWK Life Sciences Kimble™ KIMAX™ Microburets with PTFE-Plug Stopcock Available on GSA/VA Contract for Federal Government customers only.
Used for small volume titrations requiring traceable volumetric accuracy
PYREX™ Dispensing Burets with Straight Bore Product Standard PTFE Stopcock Available on GSA/VA Contract for Federal Government customers only.
For dispensing large volumes of liquids rapidly and accurately
Fisherbrand™ Automatic Burets with Stopcock
Ideal for storing standard solutions.
DWK Life Sciences Kimble™ KIMAX™ Buret Atomizer
Replacement Parts - Atomizer for 17124F Buret
Fisherbrand™ Auto-Zero Class A, Acid Buret
With reservoir and white graduations
Bel-Art™ SP Scienceware™ Automatic Self-Zeroing Glass Burets
Closed system prevents outside contamination
Eisco™ Borosilicate Burette, Straight Bore, PTFE Stopcock - 50mL cap., 0.10mL grad., Class A tolerance
Eisco Borosilicate Burette, Straight Bore, PTFE Stopcock - 50mL - Class A Burette These high quality borosilicate burettes are straight bore with interchangeable PTFE key stopcocks. Accuracy is per ISO 385, DIN 12709.
BrandTech™ BRAND™ Titrette™ Digital Bottle-Top Burets Available on GSA/VA Contract for Federal Government customers only.
Provides accuracy and precision within the tolerances of Class A glass burettes. BrandTech™ BRAND™ Titrette™ Digital Bottle-top Burets are suitable for most titrants in concentrations up to 1 mol/L. Ideal for general chemistry, wastewater treatment applications, food/beverage analysis, industrial titrations, environmental fieldwork, and more.
DWK Life Sciences Kimble™ KIMAX™ Buret Replacement Glass Barrels
Replacement glass barrels for Education-Grade Burets.
Bel-Art™ SP Scienceware™ Automatic Self-Zeroing Burets
Feature easy-to-read blue graduations against a white background
  spinner